Stopper Bags, Tyvek and HDPE, Steam Sterilization Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
0422SBSD 0508SBSD 0610SBSD 0630SBSD 0812SBSD 1035SBSD 0304SBSD 0306SBSD 0409SBSD 0412SBSD 2216SBSD 2418SBSD 2436SBSD 2440SBSD 2840SBSD 2848SBSD 1318SBSD 1414SBSD 1434SBSD 1518SBSD 1616SBSD 1632SBSD 1824SBSD 1920SBSD 2022SBSD 2026SBSD 1212SBSD 1215SBSD 1219SBSD 1238SBSD
77873-630CS 2050.05 CAD
77873-630 77873-614 77873-574 77873-654 77873-554 77873-502 77873-598 77873-608 77873-522 77873-670 77873-568 77873-510 77873-556 77873-610 77873-540 77873-656 77873-544 77873-488 77873-490 77873-652 77873-564 77873-668 77873-494 77873-612 77873-634 77873-518 77873-594 77873-500 77873-622 77873-672
Stopper Bags, Tyvek and HDPE, Steam Sterilization Indicator
Bags Sterilization Pouches
A significant upgrade over paper-based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
  • Manufactured and packaged in an ISO certified cleanroom
  • Double welded factory seals
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Heat seals closed
  • Integrated steam sterilization indicator
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Closure type: Heat seal
Opening: Cut
Delivery information:
Supplied with Certificate of Conformance.