Human Recombinant LIF (from E. coli)
Supplier: VWR International
77751-548 77751-550 77751-552 77751-554 77751-556
77751-548EA 1049.25 CAD
77751-548 77751-550 77751-552 77751-554 77751-556
Human Recombinant LIF (from E. coli)
Proteins and Peptides
Leukemia inhibitory factor (LIF) is a member of the interleukin 6 (IL-6) family that is made by a variety of adult and embryonic tissues. LIF signals through the glycoprotein 130 (gp130)/LIF receptor (LIFR) heterodimer to activate STAT3 and MAPK signaling. LIF functions during hematopoietic differentiation, neuronal cell differentiation, kidney development, and inflammatory processes. Human LIF may also be an important factor during human embryonic stem cell (hESC) self-renewal, pluripotency, and embryonic implantation.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.