Human Recombinant IL-3 (from E. coli )
Supplier:
VWR International
77751-722
77751-724
77751-726
77751-728
77751-722EA
1049.25
CAD
77751-722
77751-724
77751-726
77751-728
Human Recombinant IL-3 (from E. coli )
Proteins and Peptides
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
High quality
Low endotoxin
Affordable
Caution: For research use only.