Human Recombinant IL-3 (from E. coli)
Supplier: VWR International
77751-714 77751-716 77751-718 77751-720
77751-714EA 949.5 CAD
77751-714 77751-716 77751-718 77751-720
Human Recombinant IL-3 (from E. coli)
Proteins and Peptides
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.