Human Recombinant IL-32 A (from E. coli)
Supplier: VWR International
77750-484 77750-486 77750-488 77750-490
77750-484EA 1464.15 CAD
77750-484 77750-486 77750-488 77750-490
Human Recombinant IL-32 A (from E. coli)
Proteins and Peptides
Interleukin 32 alpha (IL-32 α) is one of six known splice variants of the IL-32 gene. IL-32 α induces the macrophage production of inflammatory cytokines, such as interleukin 8 (IL-8), tumor necrosis factor-alpha (TNF-α), and macrophage inflammatory protein 2 (MIP-2). IL-32 α expression is increased after the activation of T cells, natural killer (NK) cells, and interferon gamma-treated epithelial cells.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.