Human Recombinant FGF-21 (from E. coli)
Supplier: VWR International
77750-442 77750-444 77750-446 77750-448 77750-450
77750-442EA 949.5 CAD
77750-442 77750-444 77750-446 77750-448 77750-450
Human Recombinant FGF-21 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 21 (FGF-21) is an endocrine hormone that regulates energy homeostasis and exerts cardioprotective functions during heart injury. FGF-21 is expressed in the liver, pancreas, heart, and adipose tissues. FGF-21 signaling is activated through the FGFR1c receptor and β-Klotho co-receptor. FGF-21 is an important regulator of glucose uptake and reduces cell apoptosis under stress conditions.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.