Mouse Recombinant IL-6 (from E. coli )
Supplier:
VWR International
77751-896
77751-898
77751-900
77751-902
77751-896EA
1352.45
CAD
77751-896
77751-898
77751-900
77751-902
Mouse Recombinant IL-6 (from E. coli )
Proteins and Peptides
Interleukin 6 (IL-6) is a cytokine that is expressed by T cells, macrophages, and muscle cells. IL-6 acts to stimulate an immune response upon infection or trauma. IL-6 has both pro-inflammatory and anti-inflammatory functions, is capable of promoting fever, and signals through the cell-surface type I cytokine receptor complex containing the IL-6Ra and gp130 chains. Human IL-6 is active on mouse and rat cells.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.