Human Recombinant VEGF-165 (from E. coli)
Supplier: VWR International
77751-494 77751-496 77751-498 77751-492
77751-494EA 379.01 CAD
77751-494 77751-496 77751-498 77751-492
Human Recombinant VEGF-165 (from E. coli)
Proteins and Peptides
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.