Human Recombinant IL-1A (from E. coli)
Supplier: VWR International
77750-722 77750-724 77750-726 77750-728
77750-722EA 1464.15 CAD
77750-722 77750-724 77750-726 77750-728
Human Recombinant IL-1A (from E. coli)
Proteins and Peptides
Interleukin 1 alpha (IL-1α) is expressed by epithelial cells, activated macrophages, neutrophils, and endothelial cells to regulate immune responses. IL-1α signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter.
  • High quality
  • Low endotoxin
  • Affordable
IL-1α and the independently regulated IL-1β protein have overlapping proinflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
Caution: For research use only.