Human Recombinant EBI3 (from E. coli)
Supplier: VWR International
77750-596 77750-598 77750-600 77750-602 77750-604
77750-596EA 1464.15 CAD
77750-596 77750-598 77750-600 77750-602 77750-604
Human Recombinant EBI3 (from E. coli)
Proteins and Peptides
Epstein-Barr virus induced gene-3 (EBI3) is a secreted glycoprotein belonging to the hematopoietin receptor family related to the p40 subunit of interleukin 12 (IL-12). EBI3 expression is induced in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form interleukin 27 (IL-27), and with p35 to form interleukin 35 (IL-35). Both IL-27 and IL-35 have anti-inflammatory and regulatory activity.     
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.