Human Recombinant FGF-BASIC 154 (from E. coli)
Supplier: VWR International
77750-414 77750-416 77750-418 77750-420 77750-412
77750-414EA 151.6 CAD
77750-414 77750-416 77750-418 77750-420 77750-412
Human Recombinant FGF-BASIC 154 (from E. coli)
Proteins and Peptides
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state.
  • High quality
  • Low endotoxin
  • Affordable
Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.
Caution: For research use only.