Zero Waste Box™ Eyewear Recycling Boxes
Supplier: TERRACYCLE CANADA ULC CA
CS9-L CS9-M CS9-S
CA77867-948EA 538 CAD
CA77867-948 CA77867-950 CA77867-952
Zero Waste Box™ Eyewear Recycling Boxes
Boxes Waste Disposal Boxes
This easy-to-use recycling program provides a complete waste collection, shipment, and recycling solution for broken spectacles, safety glasses, sunglasses, glasses, ski goggles, and swim goggles.
  • Reduce landfill waste
  • Support your company's sustainability goals
  • Each collection box can be placed at convenient locations throughout the lab or facility
  • When full, simply place the collection box in a designated shipping area
  • Each box has a pre-paid UPS shipping label affixed
This program offers a convenient opportunity to recycle various types of waste found in most labs. Eyewear cases, contacts, eyewear cleaning products, contact solution, eyeglass chains and wipes, and any item contaminated with hazardous, flammable, or infectious substances are not accepted in this box.
Each sturdy collection box is made of double-corrugated cardboard. Boxes are supplied with simple assembly instructions and a pre-affixed return shipping label.