Brady® SPC® Absorbents Centurion™ Flood Barriers
Supplier: Brady Worldwide
177771 177772 177773
77775-286CS 884.78 CAD
77775-286 77775-288 77775-290
Brady® SPC® Absorbents Centurion™ Flood Barriers
Spill Control Materials
Transform flood readiness and maximize asset protection. Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Barriers.
  • Class: fresh water
  • Polypropylene fill material
  • Lint-free material texture
Centurion™ freshwater-swelled barriers contain, control, and divert flood water. Absorbent-filled design swells to full size on contact with water in 10 minutes. Flat and lightweight when dry for space-saving storage and easy positioning before first use. Easier and cleaner than sandbags. Built-in wedge prevents rolling . To divert flowing water, offset each downstream barrier toward dry side and slightly behind upstream barrier. To stack barriers, place wedge on top of wedge with two barriers, then place a third barrier on top in the middle. Cannot be used with salt water.