Brady® i75 Series R6200 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176759 176760 176761 176762
77775-036RL 0 CAD
77775-036 77775-038 77775-040 77775-042
Brady® i75 Series R6200 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbon ink wind direction: Ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6200 Series resin formulation offers superior abrasion, chemical, and heat resistance when printed on the appropriate label material from Brady®.
Recommended for printing on glossy, smooth, or semi-matte label stock, such as B-498 Repositionable Vinyl Cloth and B-486 Metallized Polyester. Combine Brady label materials with their recommended ribbon series for superior legend performance.