Brady® i75 Series R4300 Ribbons for the i7500 Label Printer
Supplier: Brady Worldwide
176746 176747 176748 176749
77775-016RL 0 CAD
77775-016 77775-018 77775-020 77775-022
Brady® i75 Series R4300 Ribbons for the i7500 Label Printer
Printer Ribbons
i75-series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbon ink wind direction: Ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R4300 Series wax/resin formulation offers strong smear and chemical resistance, and excellent barcode read rates when printed on the appropriate Brady® label material.
Recommended for printing on paper and films that have a matte or rough surface, such as B-427 Self-Laminating Vinyl, B-342 PermaSleeve markers, and B-499 Nylon Cloth from Brady®. Combine Brady label materials with their recommended ribbon series for superior legend performance.