Brady® i75 R6800 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176774 176775 176776
77775-066RL 0 CAD
77775-066 77775-068 77775-070
Brady® i75 R6800 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6800 Series white resin formulation designed for white printing on dark PermaSleeve® HT markers from Brady®.
Recommended for printing on dark-colored, heat-shrinkable tubing materials, such as black B-345 PermaSleeve® High Temperature Wire Markers from Brady®. Combine Brady label materials with their recommended ribbon series for superior legend performance.