Brady® i75 R6600 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176767 176768 176769 176770
77775-052RL 0 CAD
77775-052 77775-054 77775-056 77775-058
Brady® i75 R6600 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6600 Series resin formulation offers diesel resistance when printing on PermaSleeve® markers from Brady®.
Recommended for printing on heat-shrinkable tubing materials from Brady®, including material numbers B-342, B-7641, B-7642, B-7646, and B-7647. Combine Brady label materials with their recommended ribbon series for superior legend performance.