Extended Length Pipette Tips Empty Rack for LfTS Reload Systems, Non Sterile
Supplier: VistaLab
4080-1040 4080-2040 4080-3040
77873-986CS 82.16 CAD
77873-986 77873-990 77873-996
Extended Length Pipette Tips Empty Rack for LfTS Reload Systems, Non Sterile
Pipette Tips
Empty LfTS tip racks available without a wafer to facilitate refills with LfTS reload systems.
  • Hinged lids are designed for easy one-handed opening, making it easier to access repeatedly
  • Lids have a stacking feature for added stability and a secure locking tab
  • Lightweight lid design prevents tipping when open
  • Polypropylene
LfTS reload system tips, wafers, and racks are fully autoclavable.