NPS® Steel Height-Adjustable Science Lab Tables, Chemical-Resistant Top, Gray Frame
Supplier: National Public Seating
SLT8-3072C SLT8-4260C SLT8-2460C SLT8-2472C SLT8-3060C SLT8-2448C SLT8-2454C
470357-710EA 1152.77 CAD
470357-710 470357-716 470357-692 470357-698 470357-704 470357-680 470357-686
NPS® Steel Height-Adjustable Science Lab Tables, Chemical-Resistant Top, Gray Frame
Furniture Tables and Workbenches
Chemical resistant table top provides the ideal surface for lab experiments. NPS® steel leg science tables are the best choice for your industrial-grade table needs.
  • Steel legs adjust smoothly and easily in 1” hole increments – from 22.25 to 37.25” height
  • Leg insert built with 1⅓" square tubing 16 gauge steel frame
  • 11 gauge steel brackets
  • Legs built with 'tamper - free' hex head screws and lock nuts
  • 500 lbs. weight capacity
NPS® Steel Science Tables are built to last and made for the toughest environments. 16 gauge steel frame apron and 14 gauge legs make this table virtually indestructible. The 11 gauge steel tabletop support tabs and corner brackets fuse as one with four heavy-duty tamper-free Loctite hex screws. The adjustable height steel legs feature a supreme dual connection. The (12 gauge) leg-base is fastened to the tabletop with four heavy-duty screws which gets inserted into the frame using a strong welded hangar bolt. Legs connect to the frame corner brackets with industrial-grade nylon-insert lock nuts. The legs adjust in 1" increments and are locked firmly in place with two K-9 external tooth lock washers and heavy duty tamper-free hex screws. All legs come standard with simple-adjust glides with optional casters.
Certifications:
ANSI-BIFMA Certified; MAS Certified Green.
Ordering information:
Ten-year warranty. Seven-day lead time.