NPS® Steel Fixed-Height Science Lab Tables, Phenolic Top, Gray Frame
Supplier: National Public Seating
SLT9-2454P SLT9-2448P SLT9-3072P SLT9-2460P SLT9-2472P SLT9-3060P
470357-728EA 1502.5 CAD
470357-728 470357-722 470357-752 470357-734 470357-740 470357-746
NPS® Steel Fixed-Height Science Lab Tables, Phenolic Top, Gray Frame
Furniture Tables and Workbenches
Phenolic table top provides the ideal surface for lab experiments. NPS® steel leg science tables are the best choice for your industrial-grade table needs.
  • 14 gauge steel tubing
  • Leveling glide and caster option
  • 500 lbs. weight capacity
NPS® Steel science tables are built to last and made for the toughest environments. 16 gauge steel frame apron and 14 gauge legs make this table virtually indestructible. The 11 gauge steel tabletop support tabs and corner brackets fuse as one with 4 heavy-duty tamper-free Loctite hex screws. The steel legs feature a
More Product Information