Storage Racks for Ultra-Low Temperature Freezers
Supplier: Thermo Fisher Scientific
1950592 1950727 1950520 920090 1950592 920091 1950520
89200-162EA 410.61 CAD
89200-162 89200-186 89200-200 97018-458 CA89200-162 CA89200-186 CA89200-200
Storage Racks for Ultra-Low Temperature Freezers
Racks Cryogenic Box Racks
Cryobox storage racks makes organizing your ultra-low temperature freezer samples easy.
  • Sliding drawer racks
  • Adjustable side access box racks
  • Stores 5 cm (2") and 7.6 cm (3") cryo boxes and microplates
Certifications:
cULus and CE.