NPS® Steel Fixed-Height Science Lab Tables, HPL Top, Black Frame
Supplier: National Public Seating
SLT7-3060H SLT7-3072H SLT7-2454H SLT7-2460H SLT7-2472H SLT7-2448H
470357-668EA 1058.21 CAD
470357-668 470357-674 470357-650 470357-656 470357-662 470357-644
NPS® Steel Fixed-Height Science Lab Tables, HPL Top, Black Frame
Furniture Tables and Workbenches
High pressure laminate table top provides the ideal surface for lab experiments. NPS® Steel leg science tables are the best choice for your industrial-grade table needs.
  • 14-gauge steel tubing
  • Leveling glide and caster option
  • 500 lbs. weight capacity
NPS® Steel science tables are built to last and made for the toughest environments. 16 gauge steel frame apron and 14 gauge legs make this table virtually indestructible. The 11 gauge steel tabletop support tabs and corner brackets fuse as one with four heavy-duty tamper-free Loctite hex screws. The steel legs feature a supreme dual connection. The (12 gauge) leg-base is fastened to the tabletop with four heavy-duty screws which gets inserted into the frame using a strong welded hangar bolt. Legs connect to the frame corner brackets with industrial-grade nylon-insert lock nuts. All legs come standard with simple-adjust glides with optional caster/gusset kits.
Certifications:
ANSI-BIFMA Certified; MAS Certified Green.
Ordering information:
Ten-year warranty. Seven-day lead time.