Hanna pH Buffer Sachets
Supplier: HANNA INSTRUMENTS CANADA CA
HI70004P
HI70007P
CA470357-556CS
42.5
CAD
CA470357-556
CA470357-552
Hanna pH Buffer Sachets
Buffers
pH Calibration Buffers
This buffer value is widely used in water purification plants, in the food industry, and wherever the pH is expected to be slightly acidic.
Basic buffers that suit most users
- Certificate of Analysis (COA) on request
- Accuracy of ±0.01 pH at 25 °C
- Traceable to NIST standard
Each sealed sachet holds just the right amount of solution making every sachet as fresh as the day it was packaged. Perfect for use with testers and portable meters. Multiple resolutions for better accuracy.