Thermal Cut-Resistant Industrial Gloves with Paramide/Polyester Liner and Nitrile Dots
Supplier: MAPA
34720021 34720027 34720029
77868-184CS 1717.78 CAD
77868-184 77868-186 77868-188
Thermal Cut-Resistant Industrial Gloves with Paramide/Polyester Liner and Nitrile Dots
Gloves
Dexterity and resistance to cuts for optimized thermal protection.
  • Excellent dexterity
  • Exceptional grip
  • Thermal protection
  • ANSI cut level A2
  • Knit wrist
These gloves are designed for use in industrial environments.