Grip and Proof ³/₄ Coated Nitrile Industrial Gloves with HDPE Liner, Optipack + Recycled
Supplier: MAPA
34599050 34599051 34599057 34599058 34599059
77868-168CS 843.09 CAD
77868-168 77868-170 77868-172 77868-174 77868-176
Grip and Proof ³/₄ Coated Nitrile Industrial Gloves with HDPE Liner, Optipack + Recycled
Gloves
Gloves provide cut, grip, and skin protection up to the knuckle in oily environments.
  • Exceptional grip
  • Unbeatable fingertip sensitivity
  • Liquid-proof
  • ANSI cut level A2
  • 13 gauge HDPE liner
  • Knit wrist
These gloves are designed for use in industrial environments.