Grip and Proof Palm Coated Nitrile Industrial Gloves with HDPE Liner, Optipack + Recycled
Supplier: MAPA
34580031 34580036 34580037 34580038 34580039 34580030
77868-158CS 758.4 CAD
77868-158 77868-160 77868-162 77868-164 77868-166 77868-156
Grip and Proof Palm Coated Nitrile Industrial Gloves with HDPE Liner, Optipack + Recycled
Gloves
Cut, grip, and skin protection for oily environments.
  • Exceptional grip
  • Liquid-proof
  • ANSI cut level A2
  • 13 gauge HDPE liner
  • Knit wrist
These gloves are designed for use in industrial environments.