Saturix 3300SE Polyester Knit Dry Cleanroom Wipes, FG Cleanwipes
Supplier: FG CLEAN WIPES TE
7-3300SE-44L-S00 7-3300SE-99L-S00 7-3300SE-BBL-S00
77786-126CS 730.75 CAD
77786-126 77786-128 77786-130
Saturix 3300SE Polyester Knit Dry Cleanroom Wipes, FG Cleanwipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
FG Cleanroom Saturix 3300SE wipes provide exceptional cleanliness, strength, and absorbency. Made from 100% continuous filament polyester, these low-lint, durable wipes are ideal for critical environments, ensuring minimal particle release and contamination control for precision cleaning in cleanrooms, labs, and sensitive manufacturing applications.
  • 100% polyester knit
  • Sealed edges
  • Lightweight construction, 125 gsm
  • Double bagged in heavy mil, poly cleanroom packaging
  • Processed and packed in an ISO 4 cleanroom
  • Economy and performance
  • Low NVRS, fibers, ions,
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable