Edge® 48-939 Medium-Duty Cut Protection Industrial Gloves, Ansell
Supplier: Ansell Healthcare
48939060 48939070 48939080 48939090 48939100 48939110 48939120
77867-828CS 1645.16 CAD
77867-828 77867-830 77867-832 77867-834 77867-836 77867-838 77867-840
Edge® 48-939 Medium-Duty Cut Protection Industrial Gloves, Ansell
Gloves
Economical solution for medium-duty applications requiring high cut resistance, grip, and oil-repelling performance in oily environments.
  • NBR coated finish offers high grip in oily environments
  • Gauntlet design provides a larger oil repellent area, and easy donning and doffing
  • Thermal resistance allows the ability to confidently handle hot components
Featuring a fully NBR-coated seamless liner with a rough palm, Edge® 48-939 protective gloves offer a highly secure grip in oily environments, while protecting skin from injuries and oil contamination. The gauntlet design extends the glove's oil repellent area, and makes donning and doffing easy. The Edge® 48-939 gloves offer a high level of abrasion resistance and cut protection. In addition, the gloves provide EN 407 level 1 contact heat protection, making them ideal for workers handling hot equipment, parts, and objects.
Certifications:
ANSI/ISEA 105-2024 ABR level 6, ANSI/ISEA 105-2024 cut level A4, ANSI/ISEA 105-2024 PUN level 4, EN 407:2020 , EN ISO 21420:2020, EN 388:2016 +A1:2018.
Packaging:
Gloves are packaged 12 pairs/bag, 144 pairs/carton.