Precision Clean II Tubing Rollstock, PPC Flexible Packaging
Supplier: Fisher Container
STCK0058-002 STCK0059-002 STCK0060-003
77763-010EA 774.6 CAD
77763-010 77763-012 77763-014
Precision Clean II Tubing Rollstock, PPC Flexible Packaging
Bags Roll Stock
Precision II Tubing Rollstock offers high-quality, customizable packaging solutions designed for industries requiring durability and precision. Engineered for secure sealing and reliable performance, this rollstock is ideal for applications in healthcare, food, and industrial sectors. It ensures efficient production while maintaining product integrity and compliance with industry standards.
  • Manufactured in an ISO Class 4 Cleanroom
  • Manufactured from ultrapure clean low density polyethylene resin
  • Customizable sizing: Flexible rollstock allows users to create
More Product Information