VWR® Fume Hood Work Surface
Supplier: VWR International
VF8-0206-RF VF8-0206-RR VF8-0208-LF VF8-0208-LR VF8-0208-RF VF8-0208-RR VF8-0204-LF VF8-0204-LR VF8-0204-RF VF8-0204-RR VF8-0205-LF VF8-0205-LR VF8-0205-RF VF8-0205-RR VF8-0206-LF VF8-0206-LR
VFHWT6-RFEA 2468.2 CAD
VFHWT6-RF VFHWT6-RR VFHWT8-LF VFHWT8-LR VFHWT8-RF VFHWT8-RR VFHWT4-LF VFHWT4-LR VFHWT4-RF VFHWT4-RR VFHWT5-LF VFHWT5-LR VFHWT5-RF VFHWT5-RR VFHWT6-LF VFHWT6-LR
VWR® Fume Hood Work Surface
Fume Hoods and Enclosures Chemical Fume Hoods
Chemical resistant epoxy resin work surface designed specifically for the VWR® fume hood.
Delivery information:
Ships palletised.
Packaging:
Palletised with bandings and edge guards.
Caution:
Work surfaces are very heavy. Advise certified installers provide installation.