VWR® 950 Benchtop pH/Ion Meters
Supplier: SCHULER SCIENTIFIC
VWR-1950CL VWR-I950 77619-062 VWR-I950BF VWR-I950BR VWR-I950CA VWR-I950RC VWR-I950CLO VWR-I950CU VWR-I950F VWR-I950I VWR-I950K VWR-I950NA 77619-082 VWR-I950NH3 VWR-I950NO VWR-I950PB
77618-924EA 3686.76 CAD
77618-924 77619-060 77619-062 77619-064 77619-066 77619-068 77619-090 77619-070 77619-072 77619-074 77619-076 77619-078 77619-080 77619-082 77619-084 77619-086 77619-088
VWR® 950 Benchtop pH/Ion Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-I950 meter provides accurate and reliable measurements of pH, mV, ion concentration, and pX.
  • Color Touchscreen (7.0")
  • 1 to 6 point pH calibration with standard recognition, 1 to 6 point Ion calibration
  • Selectable pH buffer groups including NIST, DIN, GB, USA
  • Built-in ion methods including F-, Br-, NO3-, and Ag+
  • Ability for user to define ion measurement method
  • Automatic or Manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 1000 sets (GLP-compliant)
  • Automatic electrode
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable