Abgene™ Plate Covers, Thermo Scientific™
Supplier: Thermo Fisher Scientific
AB-0751 AB-0755
77753-656CS 161.76 CAD
77753-656 77753-668
Abgene™ Plate Covers, Thermo Scientific™
Lids Microplate Lids
Abgene™ plate covers provide effective sealing options for a variety of applications and can be used with 96- and 384-well Abgene™ plates. The covers are made of polypropylene or polystyrene, and are either ridged or non-ridged.
  • For use with 96- and 384-well Abgene™ plates
  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
All Abgene™ plate covers are hDNA-free, DNase-/RNase-free, and PCR inhibitor-free.