Infrared Thermometer Guns, 12:1/932 ºF
Supplier: Sper Scientific
800112 800112C
470357-592EA 47.87 CAD
470357-592 470357-590
Infrared Thermometer Guns, 12:1/932 ºF
Thermometers Digital Thermometers
Accurate IR at a reasonable price.
  • Backlit LCD
  • Laser guide
  • Min./max.
  • Audible high/low alarms
  • Memory
IR thermometers are used to safely measure the temperature in: Inaccessible, hot or hazardous objects in electrical, hazmat, HVAC, and other fields; moving machinery in factories, automobiles and elsewhere; electrical troubleshooting HVAC energy audits; avoiding contamination in the food, beverage and pharmaceutical industries.
Ordering information:
Certification is a NIST traceable certificate showing compliance with the published specifications for the product.