Platinum® Pro Sequencing Solutions, Quantum-Si
Supplier: QUANTUM-SI
910-00012-02 910-00038-00 910-00039-00
77778-308EA 1580 CAD
77778-308 77778-310 77778-312
Platinum® Pro Sequencing Solutions, Quantum-Si
Nucleic Acid Reagents Next Generation Sequencing Reagents
  • Prepare proteins for single-molecule sequencing
  • Sequence proteins with single-amino acid resolution
  • Analyze protein sequencing data with ease
How it Works: NGPS identifies proteins using unique kinetic signatures based on the binding kinetics of amino acid recognition events. Library preparation and sequencing kits contain reagents to digest and functionalize proteins and immobilize peptides on sequencing chips.
Platinum® sequences individual peptides by capturing the florescent signal from each N-terminal amino acid (NAA) binding event. Aminopeptidases cleave each N-terminal amino acid, exposing the next for recognition, and the process repeats until the whole peptide is sequenced.