Human IL-1 alpha ELISA Set (without plates)
Supplier: Abcam
AB47350-15X96TESTS AB47350-5X96TESTS AB47350-10X96TESTS
ABCA_AB47350-15X96EA 2316.28 CAD
ABCA_AB47350-15X96 ABCA_AB47350-5X96T ABCA_AB47350-10X96
Human IL-1 alpha ELISA Set (without plates)
Assays ELISAs
Human IL-1 alpha ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-1 alpha ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.