Human IL-1 beta ELISPOT Set
Supplier: Abcam
AB46578-10X96TESTS AB46578-5X96TESTS AB46578-20X96TESTS AB46578-15X96TESTS
ABCA_AB46578-10X96EA 3058.88 CAD
ABCA_AB46578-10X96 ABCA_AB46578-5X96T ABCA_AB46578-20X96 ABCA_AB46578-15X96
Human IL-1 beta ELISPOT Set
Assays Immunological Assays
Human IL-1 beta ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-1 beta production in a single cell suspension.