Human IL-23 ELISA Kit
Supplier: Abcam
AB64708-2X96TESTS AB64708-1X96TESTS
ABCA_AB64708-2X96TEA 2287.84 CAD
ABCA_AB64708-2X96T ABCA_AB64708-1X96T
Human IL-23 ELISA Kit
Assays ELISAs
Human IL-23 ELISA Kit is a sandwich (quantitative) ELISA kit for the measurement of Human IL-23 in Human in plasma, cell culture supernatant, serum samples.