TMB ELISA Substrate (Slowest Kinetic Rate)
Supplier: Abcam
AB171527-100ML AB171527-1000ML
ABCA_AB171527-100MEA 178.54 CAD
ABCA_AB171527-100M ABCA_AB171527-1000
TMB ELISA Substrate (Slowest Kinetic Rate)
Assays ELISAs
Abcam's TMB ELISA Substrate (Slowest Kinetic Rate) contains 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer.