Rat TNF alpha ELISA Kit
Supplier: Abcam
AB236712-1X96TESTS AB236712-10X96TESTS
ABCA_AB236712-1X96EA 1475.72 CAD
ABCA_AB236712-1X96 ABCA_AB236712-10X9
Rat TNF alpha ELISA Kit
Assays ELISAs
Rat TNF alpha ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat TNF alpha with a sensitivity of 1.04 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 60 citations