Human CD300a ELISA Kit
Supplier: Abcam
AB317784-1X96TESTS AB317784-10X96TESTS AB317784-1X384TESTS
ABCA_AB317784-1X96EA 1385.66 CAD
ABCA_AB317784-1X96 ABCA_AB317784-10X9 ABCA_AB317784-1X38
Human CD300a ELISA Kit
Assays ELISAs
Human CD300a ELISA kit is a single-wash 90-min Simplestep used to quantify Human CD300a with a sensitivity of 4.063 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.