Human IL-7 ELISA Set (without plates)
Supplier: Abcam
AB47357-10X96TESTS AB47357-15X96TESTS AB47357-5X96TESTS
ABCA_AB47357-10X96EA 1940.24 CAD
ABCA_AB47357-10X96 ABCA_AB47357-15X96 ABCA_AB47357-5X96T
Human IL-7 ELISA Set (without plates)
Assays ELISAs
Human IL-7 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-7 ELISA Set (without plates) in Human in plasma, cell culture supernatant, serum samples.