Human Granzyme B ELISPOT Set
Supplier: Abcam
AB46581-20X96TESTS
AB46581-5X96TESTS
AB46581-10X96TESTS
AB46581-15X96TESTS
ABCA_AB46581-20X96EA
4841.12
CAD
ABCA_AB46581-20X96
ABCA_AB46581-5X96T
ABCA_AB46581-10X96
ABCA_AB46581-15X96
Human Granzyme B ELISPOT Set
Assays
Immunological Assays
Human Granzyme B ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Granzyme B production in a single cell suspension.