Mouse Pro-Collagen I alpha 1 ELISA Kit
Supplier: Abcam
AB210579-1X384TESTS AB210579-1X96TESTS
ABCA_AB210579-1X38EA 3773.04 CAD
ABCA_AB210579-1X38 ABCA_AB210579-1X96
Mouse Pro-Collagen I alpha 1 ELISA Kit
Assays ELISAs
Mouse Pro-Collagen I alpha 1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Pro-Collagen I alpha 1 with a sensitivity of 6.7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.