Mouse IL-6 ELISA Kits
Supplier: Abcam
AB222503-1X384TESTS AB222503-1X96TESTS AB222503-10X96TESTS
ABCA_AB222503-1X38EA 3773.04 CAD
ABCA_AB222503-1X38 ABCA_AB222503-1X96 ABCA_AB222503-10X9
Mouse IL-6 ELISA Kits
Assays ELISAs
Mouse IL-6 ELISA kit is a single-wash 90-min SimpleStep ELISA® used to quantification of mouse interleukin-6 (IL-6) with a sensitivity of 11.3 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric sandwich ELISA - 450nm readout; works on any plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well for higher throughput - Broad sample compatibility - works with serum, plasma, and cell culture supernatants - Cited in over 130 publications.