Human AST ELISA Kit (Aspartate Aminotransferase)
Supplier: Abcam
AB263881-1X96TESTS AB263881-1X384TESTS
ABCA_AB263881-1X96EA 1455.18 CAD
ABCA_AB263881-1X96 ABCA_AB263881-1X38
Human AST ELISA Kit (Aspartate Aminotransferase)
Assays ELISAs
Human AST ELISA Kit (Aspartate Aminotransferase) is a single-wash 90-min Simplestep used to quantify Human AST (Aspartate Aminotransferase) with a sensitivity of 43 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair .