Human IL-10 ELISA Kit (Interleukin-10)
Supplier: Abcam
AB185986-1X384TESTS AB185986-1X96TESTS
ABCA_AB185986-1X38EA 3773.04 CAD
ABCA_AB185986-1X38 ABCA_AB185986-1X96
Human IL-10 ELISA Kit (Interleukin-10)
Assays ELISAs
Human IL-10 ELISA kit (Interleukin-10) is a single-wash, 90-minute SimpleStep ELISA® assay designed for the quantification of human IL-10 with a sensitivity of 1.4 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs: 384-well for higher throughput - Broad sample compatibility - works with serum, plasma, and cell culture supernatants.