RPMI Medium 1640 without L-Glutamine
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
9160-100ML 9160-1L 9160-500ML
77975-357EA 23.12 CAD
77975-357 77975-358 77975-359
RPMI Medium 1640 without L-Glutamine
Cell Culture Media
RPMI Medium 1640 without L-Glutamine is designed for growth of human leukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures, as well as for hybridomas.
  • Animal-component free
  • Chemically defined GMP manufacture
RPMI 1640 was developed at the Roswell Park Memorial Institute in 1966 for growing normal and neoplastic peripheral blood-derived leukocytes. Additionally, it has been used to cultivate a wide variety of cells in suspension or loosely attached cells such as hybridoma cells. When supplemented with serum or serum substitutes, RPMI 1640 is the most commonly used basal medium for long-term culture of activated T-lymphocytes.
Packaging:
Store at 2 to 8 °C and protect from light
Shelf life: 36 months from date of manufacture