BalanCD™ HEK293
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91165-1L 94137-10L
77975-340EA 207.38 CAD
77975-340 77975-398
BalanCD™ HEK293
Cell Culture Media
BalanCD HEK293 medium is a versatile, chemically-defined, animal component-free medium optimized for transfection and production of viral vectors and recombinant proteins. Chemically-defined, animal component-free supplement for enhanced galactosylation. Achieve improved protein and antibody production. The alteration of protein glycoprofiles can be a time-consuming process. BalanCD Gal supplement provides an efficient method of achieving desired results by enhancing the galactosylation of proteins with a systemized, efficient feeding schedule.
  • One medium supports multiple applications including:
  • Rapid, scalable production of viral vectors for gene therapy (adeno-associated viruses (AAV), lentiviruses)
  • Transient protein expression and recombinant protein production to gram-scale yields
  • Formulated for versatility and ease-of-use:
  • Streamline workflows with one medium for growth, seeding, transfection, and production
  • Versatile and scalable protocol for use in flasks, spinner flasks, and bioreactors
  • Maximize growth and titer by adding complementary BalanCD HEK293 feed and/or anti-clumping supplement
BalanCD HEK293 is a scalable system designed to support growth and transfection of HEK293 cell lines in suspension cultures. Comprised of BalanCD HEK293 medium, BalanCD HEK293 feed and anti-clumping supplement, this highly versatile system supports a range of applications including production of viral vectors for gene therapy, transient protein expression, and recombinant protein production. BalanCD HEK293 is part of the scalable BalanCD media and supplements platform designed to provide the optimal balance between growth and production to maximize productivity from mammalian cell cultures.
The BalanCD HEK293 System is a chemically defined, animal component-free platform of media and supplement optimized for production of viral vectors and transient protein expression. The product system comprises of BalanCD HEK293 medium, BalanCD HEK293 feed, and anti-clumping supplement. BalanCD HEK293 system contains no hydrolysates, L-Glutamine, antibiotics, antimycotics, or any other undefined components, and is ready to use for suspension culture applications. BalanCD HEK293 medium contains 6 g/L glucose and BalanCD HEK293 Feed contains 40 g/L glucose. BalanCD HEK293 medium can be supplemented with BalanCD HEK293 feed for high density cell culture, or with anti-clumping supplement post-transfection to minimize cell aggregation.
Certifications: Fulfills regulatory requirements: Chemically-defined, animal-component free, Drug Master File (DMF) filed with US FDA.
Packaging: Store at 2 to 8 °C. Shelf life: 1 year (liquid) or 2 years (solid) from date of manufacture.