Ham's F12/DME High Glucose 1:1 Mixture
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
9052-1L
9052-500ML
77975-311EA
151.1
CAD
77975-311
77975-312
Ham's F12/DME High Glucose 1:1 Mixture
Cell Culture Media
Ham’s F12/DME is a 1:1 mixture of Ham’s F12 and DMEM. This extremely rich, complex medium is commonly selected as the basal medium for the cultivation and study of challenging cell lines or primitive cells, such as neural progenitors.
Animal component-free
Chemically defined GMP manufacture