PRIME-XV FreezIS DMSO-Free
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91140-100ML 91140-10ML
77975-327EA 369.4 CAD
77975-327 77975-328
PRIME-XV FreezIS DMSO-Free
Cell Culture Supplements and Additives
DMSO-free, protein-free, animal component-free, chemically defined cryopreservation solution.
  • DMSO-free, protein-free, animal component-free, chemically defined cryopreservation solution
  • Eliminate the risk of DMSO toxicity and maintain potency of human mesenchymal stem cells (MSCs), T cells, and hematopoietic stem cells (HSCs) throughout cryopreservation
  • Comparable post-thaw cell viability to solutions containing DMSO
  • Nontoxic when injected in animal models (refer to toxicological application note)
  • Enables cell preservation for short-term at −80 °C and long-term storage in liquid nitrogen to −196 °C
  • Complete, ready-to-use medium
Human MSC and PBMC (T cell) data available for short-term storage. Human HSC data is not available. Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.
Certifications: FDA, Federal, and State registered - cGMP-compliant manufacture EN ISO 13485:2016 certified, MDSAP certified Extensive QC testing including functionality, sterility (USP <71>), endotoxin (USP <85>), and mycoplasma (USP <63>), Drug Master Files (DMFs) filed with the FDA – available upon request. By purchasing this product you are agree that you will not use this product as excipient.